missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylbutyl Bromide 97.0+%, TCI America™
Supplier: TCI America P138825G
Specifications
| 4-Phenylbutyl Bromide | |
| Yellow | |
| MFCD00154988 | |
| 1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
| BrCCCCC1=CC=CC=C1 | |
| 213.12 | |
| 213.12 | |
| Liquid |
| 13633-25-5 | |
| C10H13Br | |
| 25 g | |
| XPBQQAHIVODAIC-UHFFFAOYSA-N | |
| (4-bromobutyl)benzene | |
| 259668 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 13633-25-5 | |
| 213.12 | |
| XPBQQAHIVODAIC-UHFFFAOYSA-N | |
| 259668 | |
| BrCCCCC1=CC=CC=C1 |
| C10H13Br | |
| MFCD00154988 | |
| 1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane | |
| (4-bromobutyl)benzene |
Safety and Handling
TSCA : No