missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyl Bromide 97.0+%, TCI America™

Catalog No. p-7135111
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
P13885G 5 g
P138825G 25 g
2 options

Catalog No. P13885G

Supplier: TCI America P13885G

Only null left

Chemical Identifiers

CAS 13633-25-5
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.12
MDL Number MFCD00154988
InChI Key XPBQQAHIVODAIC-UHFFFAOYSA-N
Synonym 1-bromo-4-phenylbutane, 4-bromobutyl benzene, 4-phenylbutyl bromide, 4-phenyl 1-butyl bromide, 4-phenyl-1-butyl bromide, 4-phenyl-1-bromobutane, 1-bromo-4-phenyl-butane, benzene, 4-bromobutyl, benzene, bromobutyl, 4-phenyl-1-bromo butane
PubChem CID 259668
IUPAC Name (4-bromobutyl)benzene
SMILES BrCCCCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 213.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenylbutyl Bromide