missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenylazo)diphenylamine 98.0+%, TCI America™
Supplier: TCI America P01465G
Specifications
| 4-(Phenylazo)diphenylamine | |
| 86°C | |
| MFCD00003023 | |
| 4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
| 273.34 | |
| 273.34 | |
| Crystalline Powder |
| 101-75-7 | |
| C18H15N3 | |
| 5 g | |
| VXLFYNFOITWQPM-UHFFFAOYSA-N | |
| N-phenyl-4-(2-phenyldiazen-1-yl)aniline | |
| 7575 | |
| ≥98.0% (T) |
Chemical Identifiers
| 101-75-7 | |
| 273.34 | |
| VXLFYNFOITWQPM-UHFFFAOYSA-N | |
| 7575 | |
| N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
| C18H15N3 | |
| MFCD00003023 | |
| 4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline | |
| N-phenyl-4-(2-phenyldiazen-1-yl)aniline |
Safety and Handling
RTECSNumber : JK0185000
TSCA : Yes