accessibility menu, dialog, popup

UserName

CAS RN 101-75-7

IUPAC Name: N-phenyl-4-(2-phenyldiazen-1-yl)aniline
Synonyms: N-phenyl-4-(2-phenyldiazen-1-yl)aniline
Molecular Weight (g/mol): 273.34
Molecular Formula: C18H15N3
InChi Key: VXLFYNFOITWQPM-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)