missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenylazo)diphenylamine 98.0+%, TCI America™

Catalog No. p-7134448
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01465G 5 g
P014625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01465G Supplier TCI America Supplier No. P01465G
Only null left

Chemical Identifiers

CAS 101-75-7
Molecular Formula C18H15N3
Molecular Weight (g/mol) 273.34
MDL Number MFCD00003023
InChI Key VXLFYNFOITWQPM-UHFFFAOYSA-N
Synonym 4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline
PubChem CID 7575
IUPAC Name N-phenyl-4-(2-phenyldiazen-1-yl)aniline
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 273.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Phenylazo)diphenylamine
RTECSNumber JK0185000
TSCA Yes