missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybutyric Acid 98.0+%, TCI America™

Catalog No. P128525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P128525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P128525G Supplier TCI America Supplier No. P128525G
Only null left

Chemical Identifiers

CAS 6303-58-8
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00042656
InChI Key YKYVPFIBWVQZCE-UHFFFAOYSA-N
Synonym 4-phenoxybutyric acid, butyric acid, 4-phenoxy, butanoic acid, 4-phenoxy, 4-phenoxy-butyric acid, gamma-phenoxybutyric acid, 4-phenoxy-n-butyric acid, .gamma.-phenoxybutyric acid, 4-phenoxybutyricacid, acmc-209nce, 4-phenoxy butanoic acid
PubChem CID 22741
IUPAC Name 4-phenoxybutanoic acid
SMILES OC(=O)CCCOC1=CC=CC=C1

Specifications

Melting Point 64°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxybutyric Acid
RTECSNumber ET5956300
TSCA No