accessibility menu, dialog, popup

UserName

HOOO

CAS RN 6303-58-8

IUPAC Name: 4-phenoxybutanoic acid
Synonyms: butyric acid, 4-phenoxy-
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: YKYVPFIBWVQZCE-UHFFFAOYSA-N
SMILES: OC(=O)CCCOC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results