missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenoxybenzoic Acid 98.0+%, TCI America™
$59.69 - $232.95
Chemical Identifiers
| CAS | 2215-77-2 |
|---|---|
| Molecular Formula | C13H10O3 |
| Molecular Weight (g/mol) | 214.22 |
| MDL Number | MFCD00002539 |
| InChI Key | RYAQFHLUEMJOMF-UHFFFAOYSA-N |
| Synonym | p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh |
| PubChem CID | 75182 |
| ChEBI | CHEBI:72632 |
| IUPAC Name | 4-phenoxybenzoic acid |
| SMILES | OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 |
Chemical Identifiers
| 2215-77-2 | |
| 214.22 | |
| RYAQFHLUEMJOMF-UHFFFAOYSA-N | |
| 75182 | |
| 4-phenoxybenzoic acid |
| C13H10O3 | |
| MFCD00002539 | |
| p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh | |
| CHEBI:72632 | |
| OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 |
Specifications
| 2215-77-2 | |
| White | |
| MFCD00002539 | |
| p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh | |
| OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 | |
| 214.22 | |
| CHEBI:72632 | |
| ≥98.0% (GC,T) | |
| 4-Phenoxybenzoic Acid |
| 162°C | |
| C13H10O3 | |
| 5 g | |
| RYAQFHLUEMJOMF-UHFFFAOYSA-N | |
| 4-phenoxybenzoic acid | |
| 75182 | |
| 214.22 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DH6293620
TSCA : Yes