Learn More
4-Phenoxybenzoic acid, 99%, Thermo Scientific Chemicals
$183.59 - $183.59
Chemical Identifiers
| CAS | 2215-77-2 |
|---|---|
| Molecular Formula | C13H10O3 |
| Molecular Weight (g/mol) | 214.22 |
| MDL Number | MFCD00002539 |
| InChI Key | RYAQFHLUEMJOMF-UHFFFAOYSA-N |
| Synonym | p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh |
| PubChem CID | 75182 |
| ChEBI | CHEBI:72632 |
| IUPAC Name | 4-phenoxybenzoic acid |
| SMILES | OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1799814
|
Thermo Scientific Chemicals
A1799814 |
25 g |
Each for $183.59
|
|
|||||
Description
4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Phenoxybenzoic acid can block DNA binding of the human papillomarvirus (HPV) E2 protein. It is employed as an intermediate of Sitafloxacin, which is a fluoroquinolone antibiotic that shows promise in the treatment of Buruli ulcer.
Solubility
Very faint turbidity in hot methanol.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 2215-77-2 | |
| 214.22 | |
| RYAQFHLUEMJOMF-UHFFFAOYSA-N | |
| 75182 | |
| 4-phenoxybenzoic acid |
| C13H10O3 | |
| MFCD00002539 | |
| p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh | |
| CHEBI:72632 | |
| OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 |
Specifications
| 2215-77-2 | |
| C13H10O3 | |
| 25 g | |
| p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh | |
| RYAQFHLUEMJOMF-UHFFFAOYSA-N | |
| 4-phenoxybenzoic acid | |
| 75182 | |
| 214.22 | |
| 4-Phenoxybenzoic acid |
| 160°C to 164°C | |
| MFCD00002539 | |
| 2212463 | |
| Very faint turbidity in hot methanol. | |
| OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1 | |
| 214.22 | |
| CHEBI:72632 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-682-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only