missing translation for 'onlineSavingsMsg'
Learn More

4-Phenoxybenzenesulfonyl chloride, 97%

Catalog No. AAL1766403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1766403 1 g
AAL1766406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1766403 Supplier Thermo Scientific Chemicals Supplier No. L1766403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1623-92-3 | C12H9ClO3S | 268.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1623-92-3
Molecular Formula C12H9ClO3S
Molecular Weight (g/mol) 268.71
MDL Number MFCD00625748
InChI Key QIZPONOMFWAPRR-UHFFFAOYSA-N
Synonym 4-phenoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-phenoxy, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxyphenylsulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxy benzene sulfonyl chloride, acmc-1bvtb, chloro 4-phenoxyphenyl sulfone, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride
PubChem CID 2794756
IUPAC Name 4-phenoxybenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 42°C to 45°C
Boiling Point 180°C (5 mmHg)
Quantity 1 g
UN Number UN3260
Beilstein 2852659
Sensitivity Moisture sensitive
Formula Weight 268.72
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only