missing translation for 'onlineSavingsMsg'
Learn More

4-Pentyloxyphthalonitrile 98.0+%, TCI America™

Catalog No. p-7134925
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10711G 1 g
P10715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10711G Supplier TCI America Supplier No. P10711G
Only null left

Chemical Identifiers

CAS 106943-83-3
Molecular Formula C13H14N2O
Molecular Weight (g/mol) 214.268
MDL Number MFCD00059103
InChI Key LEYRHJFOAFEIDH-UHFFFAOYSA-N
Synonym 4-Amyloxyphthalonitrile, 1,2-Dicyano-4-pentyloxybenzene
PubChem CID 537786
IUPAC Name 4-pentoxybenzene-1,2-dicarbonitrile
SMILES CCCCCOC1=CC(=C(C=C1)C#N)C#N

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 1 g
Formula Weight 214.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pentyloxyphthalonitrile
TSCA No
Recommended Storage Refrigerator