accessibility menu, dialog, popup

UserName

H3CONN

CAS RN 106943-83-3

IUPAC Name: 4-(pentyloxy)benzene-1,2-dicarbonitrile
Synonyms: 4-(pentyloxy)benzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: LEYRHJFOAFEIDH-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC(C#N)=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results