missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentyloxyphthalonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 35°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 214.27 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Pentyloxyphthalonitrile |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |