missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 4-Oxothiane | |
| 63°C | |
| C5H8OS | |
| 25 g | |
| OVRJVKCZJCNSOW-UHFFFAOYSA-N | |
| thian-4-one | |
| 66173 | |
| ≥98.0% (GC) |
| 1072-72-6 | |
| White-Yellow | |
| MFCD00006660 | |
| tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone | |
| O=C1CCSCC1 | |
| 116.18 | |
| 116.18 | |
| Crystalline Powder |
Chemical Identifiers
| 1072-72-6 | |
| 116.18 | |
| OVRJVKCZJCNSOW-UHFFFAOYSA-N | |
| 66173 | |
| O=C1CCSCC1 |
| C5H8OS | |
| MFCD00006660 | |
| tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone | |
| thian-4-one |
Safety and Handling
TSCA : No