accessibility menu, dialog, popup

UserName

CAS RN 1072-72-6

IUPAC Name: thian-4-one
Synonyms: thian-4-one
Molecular Weight (g/mol): 116.18
Molecular Formula: C5H8OS
InChi Key: OVRJVKCZJCNSOW-UHFFFAOYSA-N
SMILES: O=C1CCSCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)