missing translation for 'onlineSavingsMsg'
Learn More

4-Oxothiane 98.0+%, TCI America™

Catalog No. p-7134192
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01125G 5 g
O011225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01125G Supplier TCI America Supplier No. O01125G
Only null left

Chemical Identifiers

CAS 1072-72-6
Molecular Formula C5H8OS
Molecular Weight (g/mol) 116.18
MDL Number MFCD00006660
InChI Key OVRJVKCZJCNSOW-UHFFFAOYSA-N
Synonym tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone
PubChem CID 66173
IUPAC Name thian-4-one
SMILES O=C1CCSCC1

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 116.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Oxothiane