missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrotoluene 99.0+%, TCI America™

Catalog No. p-7133722
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N027625G 25 g
N0276500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N027625G Supplier TCI America Supplier No. N027625G
Only null left

Chemical Identifiers

CAS 99-99-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007366
InChI Key ZPTVNYMJQHSSEA-UHFFFAOYSA-N
Synonym 4-nitrotoluene, p-nitrotoluene, 4-methylnitrobenzene, 4-nitrotoluol, benzene, 1-methyl-4-nitro, toluene, p-nitro, p-methylnitrobenzene, para-nitrotoluol, nitrotoluenos, nitrotoluene, p
PubChem CID 7473
ChEBI CHEBI:35227
IUPAC Name 1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 53°C
Boiling Point 238°C
Quantity 25 g
UN Number 3446
Formula Weight 137.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrotoluene
EINECSNumber (3)-0437
RTECSNumber XT3325000
TSCA Yes