accessibility menu, dialog, popup

UserName

CAS RN 99-99-0

IUPAC Name: 1-methyl-4-nitrobenzene
Synonyms: 4-nitrotoluene P-nitrotoluene
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: ZPTVNYMJQHSSEA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)[N+]([O-])=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)