missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol, 96%

CAS: 1849-36-1 | C6H5NO2S | 155.171 g/mol

Supplier:  Thermo Scientific Chemicals H5924006

 View more versions of this product

Catalog No. AAH5924006

Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Partly soluble in water and chloroform.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Specifications

Specifications

4-Nitrothiophenol
72°C to 77°C
MFCD00007343
606924
Partly soluble in water and chloroform.
C1=CC(=CC=C1[N+](=O)[O-])S
15809
96%
1849-36-1
C6H5NO2S
5 g
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol
AXBVSRMHOPMXBA-UHFFFAOYSA-N
155.171
155.17
Technical

Chemical Identifiers

1849-36-1
155.171
AXBVSRMHOPMXBA-UHFFFAOYSA-N
15809
C6H5NO2S
MFCD00007343
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol
C1=CC(=CC=C1[N+](=O)[O-])S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-436-4

RTECSNumber : DC1850000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only