missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothiophenol, 96%

Catalog No. p-7039802
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5924006 5 g
AAH5924014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5924006 Supplier Thermo Scientific Chemicals Supplier No. H5924006

CAS: 1849-36-1 | C6H5NO2S | 155.171 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Partly soluble in water and chloroform.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1849-36-1
Molecular Formula C6H5NO2S
Molecular Weight (g/mol) 155.171
MDL Number MFCD00007343
InChI Key AXBVSRMHOPMXBA-UHFFFAOYSA-N
Synonym 4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol
PubChem CID 15809
SMILES C1=CC(=CC=C1[N+](=O)[O-])S

Specifications

Melting Point 72°C to 77°C
Quantity 5 g
Beilstein 606924
Solubility Information Partly soluble in water and chloroform.
Formula Weight 155.17
Percent Purity 96%
Grade Technical
Chemical Name or Material 4-Nitrothiophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-436-4
RTECSNumber DC1850000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only