missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothioanisole 98.0+%, TCI America™

Catalog No. N07615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07615G 5 g
N076125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07615G Supplier TCI America Supplier No. N07615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-57-5
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
InChI Key NEZGPRYOJVPJKL-UHFFFAOYSA-N
Synonym 4-nitrothioanisole, 1-methylthio-4-nitrobenzene, methyl 4-nitrophenyl sulfide, 1-methylsulfanyl-4-nitrobenzene, benzene, 1-methylthio-4-nitro, methyl 4-nitrophenyl sulphide, 1-methylsulfanyl-4-nitro-benzene, pubchem10631, 4-methylthio nitrobenzene
PubChem CID 96109
IUPAC Name 1-methylsulfanyl-4-nitrobenzene
SMILES CSC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 71°C
Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 169.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrothioanisole