accessibility menu, dialog, popup

UserName

CAS RN 701-57-5

IUPAC Name: 1-(methylsulfanyl)-4-nitrobenzene
Synonyms: 1-(methylsulfanyl)-4-nitrobenzene
Molecular Weight (g/mol): 169.2
Molecular Formula: C7H7NO2S
InChi Key: NEZGPRYOJVPJKL-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)