missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrothioanisole, 98%

Catalog No. AAB2033906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2033906 5 g
AAB2033914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2033906 Supplier Thermo Scientific Chemicals Supplier No. B2033906

CAS: 701-57-5 | C7H7NO2S | 169.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 701-57-5
Molecular Formula C7H7NO2S
Molecular Weight (g/mol) 169.198
MDL Number MFCD00010868
InChI Key NEZGPRYOJVPJKL-UHFFFAOYSA-N
Synonym 4-nitrothioanisole, 1-methylthio-4-nitrobenzene, methyl 4-nitrophenyl sulfide, 1-methylsulfanyl-4-nitrobenzene, benzene, 1-methylthio-4-nitro, methyl 4-nitrophenyl sulphide, 1-methylsulfanyl-4-nitro-benzene, pubchem10631, 4-methylthio nitrobenzene
PubChem CID 96109
IUPAC Name 1-methylsulfanyl-4-nitrobenzene
SMILES CSC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 66°C to 72°C
Quantity 5 g
Beilstein 1817928
Formula Weight 169.2
Percent Purity 98%
Chemical Name or Material 4-Nitrothioanisole
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only