missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalonitrile 98.0+%, TCI America™

$126.76 - $447.38

Chemical Identifiers

CAS 31643-49-9
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00040301
InChI Key NTZMSBAAHBICLE-UHFFFAOYSA-N
Synonym 4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
PubChem CID 97443
IUPAC Name 4-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N052425G
View Documents
TCI America
N052425G
25 g
Each for $126.76
Only null left
 
N0524100G
View Documents
TCI America
N0524100G
100 g
Each for $447.38
Only null left
 
Description

Description

Specifications

Chemical Identifiers

31643-49-9
173.13
NTZMSBAAHBICLE-UHFFFAOYSA-N
97443
[O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
C8H3N3O2
MFCD00040301
4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
4-nitrobenzene-1,2-dicarbonitrile

Specifications

31643-49-9
C8H3N3O2
25 g
NTZMSBAAHBICLE-UHFFFAOYSA-N
4-nitrobenzene-1,2-dicarbonitrile
97443
≥98.0% (GC)
4-Nitrophthalonitrile
142°C
MFCD00040301
4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
[O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N
173.13
173.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TI8576000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'