missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic anhydride, tech. 90%

Catalog No. AAL0802006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0802006 5 g
AAL0802014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0802006 Supplier Thermo Scientific Chemicals Supplier No. L0802006

CAS: 5466-84-2 | C8H3NO5 | 193.114 g/mol

4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophthalic anhydride is used an an intermediate for the synthesis of a benzimidazole PARP inhibitor I (succinate salt) (ABT-472).

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents. Incompatible with water, bases, strong acids and oxidizing agents.

Chemical Identifiers

CAS 5466-84-2
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.114
MDL Number MFCD00005922
InChI Key MMVIDXVHQANYAE-UHFFFAOYSA-N
Synonym 4-nitrophthalic anhydride, 5-nitroisobenzofuran-1,3-dione, phthalic anhydride, 4-nitro, 1,3-isobenzofurandione, 5-nitro, 4-nitrophthalic acid anhydride, 5-nitrophthalic anhydride, unii-sqn32ua63n, ccris 4684, 5-nitro-1,3-isobenzofurandione, sqn32ua63n
PubChem CID 230976
IUPAC Name 5-nitro-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)OC2=O

Specifications

Melting Point 116°C to 121°C
Quantity 5 g
Beilstein 179682
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 193.11
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Nitrophthalic anhydride
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-776-2
RTECSNumber TI3328000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only