accessibility menu, dialog, popup

UserName

ONOOOO

CAS RN 5466-84-2

IUPAC Name: 5-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-nitro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 193.11
Molecular Formula: C8H3NO5
InChi Key: MMVIDXVHQANYAE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)