missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic Anhydride 98.0+%, TCI America™

Catalog No. N024625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N024625G 25 g
N0246100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N024625G Supplier TCI America Supplier No. N024625G

Chemical Identifiers

CAS 5466-84-2
Molecular Formula C8H3NO5
Molecular Weight (g/mol) 193.114
MDL Number MFCD00005922
InChI Key MMVIDXVHQANYAE-UHFFFAOYSA-N
Synonym 4-nitrophthalic anhydride, 5-nitroisobenzofuran-1,3-dione, phthalic anhydride, 4-nitro, 1,3-isobenzofurandione, 5-nitro, 4-nitrophthalic acid anhydride, 5-nitrophthalic anhydride, unii-sqn32ua63n, ccris 4684, 5-nitro-1,3-isobenzofurandione, sqn32ua63n
PubChem CID 230976
IUPAC Name 5-nitro-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)OC2=O

Specifications

Melting Point 120°C
Color Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 193.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalic Anhydride
RTECSNumber TI3328000
TSCA Yes