missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetonitrile, 98%

Catalog No. p-7022179
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1482118 50 g
AAA1482130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1482118 Supplier Thermo Scientific Chemicals Supplier No. A1482118
Only null left

CAS: 555-21-5 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 555-21-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00007372
InChI Key PXNJGLAVKOXITN-UHFFFAOYSA-N
Synonym 4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
PubChem CID 68386
IUPAC Name 2-(4-nitrophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)[N+](=O)[O-]

Specifications

Melting Point 114°C to 117°C
Boiling Point 195°C to 197°C (12 mmHg)
Quantity 50 g
UN Number UN3439
Beilstein 388442
Merck Index 14,6590
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 209-085-0
RTECSNumber AM1250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only