accessibility menu, dialog, popup

UserName

ONON

CAS RN 555-21-5

IUPAC Name: 2-(4-nitrophenyl)acetonitrile
Synonyms: benzeneacetonitrile, 4-nitro-
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: PXNJGLAVKOXITN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC#N)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)