missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrophenyl)urea 98.0+%, TCI America™

Catalog No. p-7134062
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09801G 1 g
N09805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09801G Supplier TCI America Supplier No. N09801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 556-10-5
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD09040599
InChI Key LXXTVGKSGJADFU-UHFFFAOYSA-N
PubChem CID 313520
IUPAC Name (4-nitrophenyl)urea
SMILES C1=CC(=CC=C1NC(=O)N)[N+](=O)[O-]

Specifications

Melting Point 217°C
Color Yellow
Quantity 1 g
Formula Weight 181.15
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Nitrophenyl)urea