missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phenyl sulfide, 98%

Catalog No. AAA1727814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1727814 25 g
AAA1727806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1727814 Supplier Thermo Scientific Chemicals Supplier No. A1727814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 952-97-6 | C12H9NO2S | 231.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 952-97-6
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.27
MDL Number MFCD00024700
InChI Key RJCBYBQJVXVVKB-UHFFFAOYSA-N
Synonym 4-nitrophenyl phenyl sulfide, 4-nitrodiphenyl sulfide, 1-nitro-4-phenylsulfanyl benzene, p-nitrodiphenyl sulfide, p-nitrophenyl phenyl sulfide, 4-nitrodiphenyl thioether, 1-nitro-4-phenylthio benzene, phenyl p-nitrophenylsulfide, benzene, 1-nitro-4-phenylthio, sulfide, p-nitrophenyl phenyl
PubChem CID 13720
IUPAC Name 1-nitro-4-phenylsulfanylbenzene
SMILES [O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1

Specifications

Melting Point 54°C to 57°C
Quantity 25 g
Formula Weight 231.27
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl phenyl sulfide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-462-5
RTECSNumber WQ5620000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only