CAS RN 952-97-6
IUPAC Name:
1-nitro-4-(phenylsulfanyl)benzene
Synonyms:
1-nitro-4-(phenylsulfanyl)benzene
Molecular Weight (g/mol):
231.27
Molecular Formula:
C12H9NO2S
InChi Key:
RJCBYBQJVXVVKB-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Nitrophenyl phenyl sulfide, 98%
4-Nitrophenyl phenyl sulfide, 98%
CAS: 952-97-6 Molecular Formula: C12H9NO2S Molecular Weight (g/mol): 231.27 MDL Number: MFCD00024700 InChI Key: RJCBYBQJVXVVKB-UHFFFAOYSA-N Synonym: 4-nitrophenyl phenyl sulfide,4-nitrodiphenyl sulfide,1-nitro-4-phenylsulfanyl benzene,p-nitrodiphenyl sulfide,p-nitrophenyl phenyl sulfide,4-nitrodiphenyl thioether,1-nitro-4-phenylthio benzene,phenyl p-nitrophenylsulfide,benzene, 1-nitro-4-phenylthio,sulfide, p-nitrophenyl phenyl PubChem CID: 13720 IUPAC Name: 1-nitro-4-phenylsulfanylbenzene SMILES: [O-][N+](=O)C1=CC=C(SC2=CC=CC=C2)C=C1