missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isothiocyanate, 97%

CAS: 2131-61-5 | C7H4N2O2S | 180.181 g/mol

$102.74 - $972.68

Chemical Identifiers

CAS 2131-61-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD00007307
InChI Key NXHSSIGRWJENBH-UHFFFAOYSA-N
Synonym 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
PubChem CID 75044
IUPAC Name 1-isothiocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=S)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1104209
View Documents
Thermo Scientific Chemicals
L1104209
10 g
Each for $102.74
Only null left
 
AAL1104218
View Documents
Thermo Scientific Chemicals
L1104218
50 g
Each for $315.99
Only null left
 
AAL1104230
View Documents
Thermo Scientific Chemicals
L1104230
250 g
Each for $972.68
Only null left
 
Description

Description

4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

2131-61-5
180.181
NXHSSIGRWJENBH-UHFFFAOYSA-N
75044
C1=CC(=CC=C1N=C=S)[N+](=O)[O-]
C7H4N2O2S
MFCD00007307
4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
1-isothiocyanato-4-nitrobenzene

Specifications

2131-61-5
137°C to 138°C (11 mmHg)
C7H4N2O2S
10 g
640027
4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
NXHSSIGRWJENBH-UHFFFAOYSA-N
1-isothiocyanato-4-nitrobenzene
75044
97%
105°C to 110°C
Irritating
MFCD00007307
UN2923
Moisture sensitive
It hydrolyzes with water. Soluble in toluene.
C1=CC(=CC=C1N=C=S)[N+](=O)[O-]
180.181
180.19
4-Nitrophenyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c

H302+H312+H332-H315-H317-H319-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 218-359-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only