Learn More
4-Nitrophenyl isothiocyanate, 97%
CAS: 2131-61-5 | C7H4N2O2S | 180.181 g/mol
Supplier: Thermo Scientific Chemicals L1104218
Description
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
Solubility
It hydrolyzes with water. Soluble in toluene.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
| 4-Nitrophenyl isothiocyanate | |
| 105°C to 110°C | |
| Irritating | |
| MFCD00007307 | |
| UN2923 | |
| Moisture sensitive | |
| It hydrolyzes with water. Soluble in toluene. | |
| C1=CC(=CC=C1N=C=S)[N+](=O)[O-] | |
| 180.181 | |
| 180.19 |
| 2131-61-5 | |
| 137°C to 138°C (11 mmHg) | |
| C7H4N2O2S | |
| 50 g | |
| 640027 | |
| 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate | |
| NXHSSIGRWJENBH-UHFFFAOYSA-N | |
| 1-isothiocyanato-4-nitrobenzene | |
| 75044 | |
| 97% |
Chemical Identifiers
| 2131-61-5 | |
| 180.181 | |
| NXHSSIGRWJENBH-UHFFFAOYSA-N | |
| 75044 | |
| C1=CC(=CC=C1N=C=S)[N+](=O)[O-] |
| C7H4N2O2S | |
| MFCD00007307 | |
| 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate | |
| 1-isothiocyanato-4-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 218-359-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only