Learn More
4-Nitrophenyl formate, 98%
CAS: 1865-01-6 | C7H5NO4 | 167.12 g/mol
Supplier: Thermo Scientific Chemicals L0031614
Description
Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsConvenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.
Solubility
Hydrolyzes in water. Soluble in dichloromethane.
Notes
Moisture and light sensitive. Store away from water/moisture, oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark. Store under inert gas. Protect from exposure to light.
Specifications
| 4-Nitrophenyl formate | |
| 70°C to 74°C | |
| MFCD00014713 | |
| 1950897 | |
| p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc | |
| IEXRKQFZXJSHOB-UHFFFAOYSA-N | |
| (4-nitrophenyl) formate | |
| 74628 | |
| 98% |
| 1865-01-6 | |
| C7H5NO4 | |
| 25 g | |
| Light and moisture sensitive | |
| Hydrolyzes in water. Soluble in dichloromethane. | |
| C1=CC(=CC=C1[N+](=O)[O-])OC=O | |
| 167.12 | |
| 167.12 |
Chemical Identifiers
| 1865-01-6 | |
| 167.12 | |
| IEXRKQFZXJSHOB-UHFFFAOYSA-N | |
| (4-nitrophenyl) formate |
| C7H5NO4 | |
| MFCD00014713 | |
| 74628 | |
| C1=CC(=CC=C1[N+](=O)[O-])OC=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 217-473-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only