accessibility menu, dialog, popup

UserName

CAS RN 1865-01-6

IUPAC Name: 4-nitrophenyl formate
Synonyms: 4-nitrophenyl formate
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: IEXRKQFZXJSHOB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results