missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 98%

Catalog No. AAL0031606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0031606 5 g
AAL0031614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0031606 Supplier Thermo Scientific Chemicals Supplier No. L0031606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1865-01-6 | C7H5NO4 | 167.12 g/mol

Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Convenient and selective reagent for the selective formylation of terminal amino groups in such basic amino acids as ornithine or lysine and formylation of α-amino groups in peptides.

Solubility
Hydrolyzes in water. Soluble in dichloromethane.

Notes
Moisture and light sensitive. Store away from water/moisture, oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark. Store under inert gas. Protect from exposure to light.

Chemical Identifiers

CAS 1865-01-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00014713
InChI Key IEXRKQFZXJSHOB-UHFFFAOYSA-N
PubChem CID 74628
IUPAC Name (4-nitrophenyl) formate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC=O

Specifications

Melting Point 70°C to 74°C
Quantity 5 g
Beilstein 1950897
Sensitivity Light and moisture sensitive
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Solubility Information Hydrolyzes in water. Soluble in dichloromethane.
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl formate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-473-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only