missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Acetate 98.0+%, TCI America™

Supplier:  TCI America A004025G

 View more versions of this product

Catalog No. A004025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Nitrophenyl Acetate
78°C
C8H7NO4
25 g
QAUUDNIGJSLPSX-UHFFFAOYSA-N
4-nitrophenyl acetate
13243
181.15
Crystalline Powder
830-03-5
Yellow
MFCD00007326
p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
181.15
CHEBI:82635
≥98.0% (GC)

Chemical Identifiers

830-03-5
181.15
QAUUDNIGJSLPSX-UHFFFAOYSA-N
13243
4-nitrophenyl acetate
C8H7NO4
MFCD00007326
p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
CHEBI:82635
CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : AJ1150000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions