accessibility menu, dialog, popup

UserName

CAS RN 830-03-5

IUPAC Name: 4-nitrophenyl acetate
Synonyms: P-nitrophenylacetate
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: QAUUDNIGJSLPSX-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)