Learn More
4-Nitrophenol, 99%
CAS: 100-02-7 | C6H5NO3 | 139.11 g/mol
Supplier: Thermo Scientific Chemicals A143760B
Description
4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.
Solubility
Freely soluble in alcohol, chloroform, ether. Freely soluble in acetone. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
| 4-Nitrophenol | |
| 110°C to 115°C | |
| 279°C (decomposition) | |
| C6H5NO3 | |
| 1000 g | |
| 1281877 | |
| 14,6621 | |
| Freely soluble in alcohol,chloroform,ether; Freely soluble in acetone; Slightly soluble in water. | |
| OC1=CC=C(C=C1)[N+]([O-])=O | |
| 139.11 | |
| CHEBI:16836 | |
| 99% |
| 100-02-7 | |
| 1.48 | |
| 169°C (336°F) | |
| MFCD00007331 | |
| UN1663 | |
| Light sensitive | |
| p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo | |
| BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
| 4-nitrophenol | |
| 980 | |
| 139.11 |
Chemical Identifiers
| 100-02-7 | |
| 139.11 | |
| BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
| 980 | |
| 4-nitrophenol |
| C6H5NO3 | |
| MFCD00007331 | |
| p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo | |
| CHEBI:16836 | |
| OC1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
GHS H Statement
H373-H302-H312-H332
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P312-P314-P330-P363-P501c
H301-H312+H332-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
EINECSNumber : 202-811-7
RTECSNumber : SM2275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only