accessibility menu, dialog, popup

UserName

CAS RN 100-02-7

IUPAC Name: 4-nitrophenol
Synonyms: P-nitrophenol paranitrophenol
Molecular Weight (g/mol): 139.11
Molecular Formula: C6H5NO3
InChi Key: BTJIUGUIPKRLHP-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (7)
Boiling Point 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (1)