missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroguaiacol, 97%

CAS: 3251-56-7 | C7H7NO4 | 169.14 g/mol

$82.11 - $82.11

Identifiants chimiques

CAS 3251-56-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00012143
InChI Key IZLVFLOBTPURLP-UHFFFAOYSA-N
Synonym 4-nitroguaiacol, phenol, 2-methoxy-4-nitro, guaiacol, 4-nitro, 2-hydroxy-5-nitroanisole, 3-nitro-6-hydroxyanisole, 4-hydroxy-3-methoxynitrobenzene, phenol, o-methoxy-p-nitro, o-methoxy-p-nitrophenol, 2-methoxy-4-nitro-phenol, 4-nitrogualacol
PubChem CID 76738
ChEBI CHEBI:81050
IUPAC Name 2-methoxy-4-nitrophenol
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC310370050
FDS Afficher les documents
Thermo Scientific Chemicals
310370050
5 g Glass bottle
chaque for $82.11
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

3251-56-7
169.14
IZLVFLOBTPURLP-UHFFFAOYSA-N
76738
2-methoxy-4-nitrophenol
C7H7NO4
MFCD00012143
4-nitroguaiacol, phenol, 2-methoxy-4-nitro, guaiacol, 4-nitro, 2-hydroxy-5-nitroanisole, 3-nitro-6-hydroxyanisole, 4-hydroxy-3-methoxynitrobenzene, phenol, o-methoxy-p-nitro, o-methoxy-p-nitrophenol, 2-methoxy-4-nitro-phenol, 4-nitrogualacol
CHEBI:81050
COC1=C(C=CC(=C1)[N+](=O)[O-])O

Spécifications

3251-56-7
100.0
Yellow
97%
C7H7NO4
MFCD00012143
4-nitroguaiacol, phenol, 2-methoxy-4-nitro, guaiacol, 4-nitro, 2-hydroxy-5-nitroanisole, 3-nitro-6-hydroxyanisole, 4-hydroxy-3-methoxynitrobenzene, phenol, o-methoxy-p-nitro, o-methoxy-p-nitrophenol, 2-methoxy-4-nitro-phenol, 4-nitrogualacol
IZLVFLOBTPURLP-UHFFFAOYSA-N
2-methoxy-4-nitrophenol
76738
169.14
Powder
96.0
99.0°C to 104.0°C
Authentic
Glass bottle
O2NC6H3(OCH3)OH
5 g
Solubility in water: insoluble in water
COC1=C(C=CC(=C1)[N+](=O)[O-])O
169.14
CHEBI:81050
97%
4-Nitroguaiacol
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 221-839-

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only