accessibility menu, dialog, popup

UserName

CAS RN 3251-56-7

IUPAC Name: 2-methoxy-4-nitrophenol
Synonyms: 2-methoxy-4-nitrophenol
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: IZLVFLOBTPURLP-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)