missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrodiphenyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 61°C |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 215.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrodiphenyl Ether |
Safety and Handling
| RTECSNumber | KO2680000 |
| TSCA | Yes |