accessibility menu, dialog, popup

UserName

CAS RN 620-88-2

IUPAC Name: 1-nitro-4-phenoxybenzene
Synonyms: 4-nitro diphenyl ether
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: JDTMUJBWSGNMGR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results