missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrobenzyl)triphenylphosphonium bromide, 98%

CAS: 2767-70-6 | C25H21BrNO2P | 478.33 g/mol

$46.95 - $144.59

Chemical Identifiers

CAS 2767-70-6
Molecular Formula C25H21BrNO2P
Molecular Weight (g/mol) 478.33
MDL Number MFCD00032108
InChI Key IPJPTPFIJLFWLP-UHFFFAOYSA-M
Synonym 4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide
PubChem CID 197008
IUPAC Name (4-nitrophenyl)methyl-triphenylphosphanium;bromide
SMILES [Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1528806
View Documents
Thermo Scientific Chemicals
A1528806
5 g
Each for $46.95
Only null left
 
AAA1528814
View Documents
Thermo Scientific Chemicals
A1528814
25 g
Each for $144.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2767-70-6
478.33
IPJPTPFIJLFWLP-UHFFFAOYSA-M
197008
[Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
C25H21BrNO2P
MFCD00032108
4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide
(4-nitrophenyl)methyl-triphenylphosphanium;bromide

Specifications

2767-70-6
C25H21BrNO2P
5 g
IPJPTPFIJLFWLP-UHFFFAOYSA-M
(4-nitrophenyl)methyl-triphenylphosphanium;bromide
197008
98%
∼280°C (decomposition)
MFCD00032108
4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide
[Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
478.33
478.33
(4-Nitrobenzyl)triphenylphosphonium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-444-0

RTECSNumber : TA2390000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only