missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Cyanide 98.0+%, TCI America™

Catalog No. N022925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N022925G 25 g
N0229500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N022925G Supplier TCI America Supplier No. N022925G

Chemical Identifiers

CAS 555-21-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00007372
InChI Key PXNJGLAVKOXITN-UHFFFAOYSA-N
Synonym 4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
PubChem CID 68386
IUPAC Name 2-(4-nitrophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)[N+](=O)[O-]

Specifications

Melting Point 115°C
Color Yellow
Boiling Point 197°C
Quantity 25 g
UN Number 3439
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Cyanide
RTECSNumber AM1250000
TSCA Yes