missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Chloroformate 97.0+%, TCI America™

Catalog No. C107725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C107725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C107725G Supplier TCI America Supplier No. C107725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4457-32-3
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00007375
InChI Key MHSGOABISYIYKP-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloroformate, 4-nitrobenzyl carbonochloridate, carbonochloridic acid, 4-nitrophenyl methyl ester, 4-nitrobenzyl chlorocarbonate, p-nitrobenzyl chloroformate, 4-nitrocarbobenzoxychloride, 4-nitrophenyl methyl chloroformate, p-nitrobenzyloxycarbonyl chloride, 4-nitrobenzyloxycarbonyl chloride, chloroformic acid p-nitrobenzyl ester
PubChem CID 78205
IUPAC Name (4-nitrophenyl)methyl carbonochloridate
SMILES C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 230°C
Quantity 25 g
UN Number 1759
Formula Weight 215.59
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Chloroformate
EINECSNumber (3)-4629
TSCA Yes
Recommended Storage Refrigerator