missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Chloride 98.0+%, TCI America™

Catalog No. C022925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022925G 25 g
C0229250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C022925G Supplier TCI America Supplier No. C022925G

Chemical Identifiers

CAS 100-14-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007374
InChI Key KGCNHWXDPDPSBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
PubChem CID 7482
ChEBI CHEBI:87406
IUPAC Name 1-(chloromethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCl)[N+](=O)[O-]

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Chloride
EINECSNumber (3)-0449
RTECSNumber XS9093000
TSCA Yes