missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloride, 99%

Catalog No. AC161092500
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161092500 250 g Glass bottle
AC161090100 10 g Glass bottle
AC161091000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161092500 Supplier Thermo Scientific Chemicals Supplier No. 161092500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-14-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007374
InChI Key KGCNHWXDPDPSBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
PubChem CID 7482
ChEBI CHEBI:87406
IUPAC Name 1-(chloromethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCl)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 71.0°C to 74.0°C
Color Yellow
Boiling Point 112.0°C (0.6 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 250 g
Beilstein 05,329
Fieser 01,736
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,aceton and benzene
Formula Weight 171.58
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-Nitrobenzyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-822-7
RTECSNumber XS9093000
TSCA TSCA

RUO – Research Use Only